Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCDHAC2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | PCDHAC2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PCDHAC2 Polyclonal specifically detects PCDHAC2 in Human samples. It is validated for Western Blot.Specifications
PCDHAC2 | |
Polyclonal | |
Rabbit | |
Neuroscience | |
MGC71598, PCDH-ALPHA-C2, protocadherin alpha subfamily C, 2, protocadherin alpha-C2 | |
PCDHAC2 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
Q9Y5I4 | |
56134 | |
Synthetic peptides corresponding to PCDHAC2(protocadherin alpha subfamily C, 2) The peptide sequence was selected from the N terminal of PCDHAC2. Peptide sequence SPAFDQSTYRVQLREDSPPGTLVVKLNASDPDEGSNGELRYSLSSYTSDR. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title