Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDHB10 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983109
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3109 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3109 Supplier Bioss Supplier No. BS13725R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintenance of specific neuronal connections in the brain.

Specifications

Antigen PCDHB10
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PCDHB10
Gene Accession No. Q9UN67
Gene Alias PCDHB10; PCDH-BETA10; PCDH-beta-10; PCHB10; protocadherin beta 10; Protocadherin beta-10; UNQ1906/PRO4352
Gene Symbols PCDHB10
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PCDHB10.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56126
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.