Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDHB12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571679
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571679 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571679 Supplier Invitrogen™ Supplier No. PA571679
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PCDHB12 Polyclonal Antibody for Western Blot, IHC (P)

Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintenance of specific neuronal connections in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDHB12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PCDHB12
Gene Accession No. Q9Y5F1
Gene Alias Pcdh3; PCDHB12; Pcdhb5F; PCDH-BETA12; PCDH-beta-12; PcdhbL; protocadherin 3; protocadherin beta 12; protocadherin beta-12
Gene Symbols PCDHB12
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 766-795 amino acids from the C-terminal region of human PCDHB12.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56124
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.