Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDHB15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550726
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550726 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550726 Supplier Invitrogen™ Supplier No. PA550726
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total PCDHB15 protein.

Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintenance of specific neuronal connections in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDHB15
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PCDHB15
Gene Accession No. Q9Y5E8
Gene Alias PCDHB15; PCDH-BETA15; PCDH-beta-15; protocadherin beta 15; protocadherin beta-15
Gene Symbols PCDHB15
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human protocadherin beta 15.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56121
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.