Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCDHB15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584957
Description
Immunogen sequence: FLKPIFPNIV SQDSRRKSEF LE.
Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintenance of specific neuronal connections in the brain.
Specifications
PCDHB15 | |
Polyclonal | |
Unconjugated | |
PCDHB15 | |
PCDHB15; PCDH-BETA15; PCDH-beta-15; protocadherin beta 15; protocadherin beta-15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56121 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y5E8 | |
PCDHB15 | |
Recombinant protein corresponding to Human PCDHB15. Recombinant protein control fragment (Product #RP-108637). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction