Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCDHB16 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219727
Description
PCDHB16 Polyclonal specifically detects PCDHB16 in Human, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PCDHB16 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
cadherin ME1, KIAA1621PCDHbeta 16, ME1, PCDH3Xprotocadherin beta 8a, PCDHB 16, PCDHB8a, PCDH-beta-16, PCDH-BETA16, protocadherin beta 16, protocadherin beta-16, Protocadherin-3X | |
Rabbit | |
85 kDa | |
0.1 mL | |
Primary | |
Human, Mouse (Negative) | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Q9NRJ7 | |
PCDHB16 | |
Recombinant protein encompassing a sequence within the N-terminus region of human PCDHB16. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
57717 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction