Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDHB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120577
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120577 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120577 Supplier Invitrogen™ Supplier No. PA5120577
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, Raji, Mouse testis, Rat testis. Immunogen sequence: EPIRYSVLEE TESGSFVAHL AKDLGLGIGE LASRSARVLS DDDKQRLQLD RQTGDLLLRE KLDREELCGP IEPCVLHFQV FLEMPVQFFQ GELLIQDIND HSPIFPEREV LLKILENSQP GTLFPLLIAE DLDVGSNGLQ KYTISPNSHF HILTRNHSEG KKYPDLVQDK PLDREEQPEF SLTLVALDGG SPPRSGTVMV RILIMDINDN APEFVHTPYG VQVLENSPLD SPIVRVLARD IDAGNFGSVS YGLFQASDEI KQTFSINEVT GEI.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDHB4
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PCDHB4
Gene Accession No. Q9Y5E5
Gene Alias PCDHB4; Pcdhb5A; PcdhbD; PCDH-BETA4; PCDH-beta-4; protocadherin beta 4; protocadherin beta-4
Gene Symbols PCDHB4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 28-300 of human PCDHB4 (NP_061761.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291655, 56131, 93875
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.