Learn More
Invitrogen™ PCDHB6 Monoclonal Antibody (2G11)

Description
Peptide Sequence: ASLRVRDIND HAPEFPAREM LLKISEITMP GKIFPLKMAH DLDTGSNGLQ RYTISSNPHF HVLTRNRSEG RKFPELVLDK PLDREEQPQL RLTLIALDGG.
Specifications
Specifications
| Antigen | PCDHB6 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2G11 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | PCDHB6 |
| Gene Accession No. | Q9Y5E3 |
| Gene Alias | PCDHB6; PCDH-BETA6; PCDH-beta-6; POLR1E; protocadherin beta 6; protocadherin beta-6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.