Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCDHB8 Monoclonal Antibody (OTI1F6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA812703

Description
Calcium-dependent cell-adhesion protein involved in cells self-recognition and non-self discrimination. Thereby, it is involved in the establishment and maintece of specific neuronal connections in the brain.Specifications
PCDHB8 | |
Monoclonal | |
Unconjugated | |
PCDHB8 | |
PCDH-BETA8, PCDH3I | |
Mouse | |
Affinity Chromatography | |
RUO | |
56128 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI1F6 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9UN66 | |
PCDHB8 | |
Human recombit protein fragment corresponding to amino acids 29-418 of human Pcdhb8 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction