Learn More
PCDHGA1 Antibody (S159-5), Alexa Fluor™ 700, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP242207AF700
Catalog No. NB242207700
Only null left
Add to Cart
Description
Description
PCDHGA1 Monoclonal specifically detects PCDHGA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
PCDHGA1
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
PCDHGA1
Fusion protein corresponding to amino acids 808-931 (C-terminal cytoplasmic constant domain) of mouse Protocadherin-gamma-A1 that is shared by all 22 Gamma-protocadherins (A subfamily amino acids ∼807-930, B subfamily amino acids ∼789-912 and C subfamily amino acids ∼818-941). 99% identity with human (123/124 amino acids).
0.1 mL
56114
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
S159-5
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
MGC138287, MGC138289, PCDH-GAMMA-A1, protocadherin gamma subfamily A, 1, protocadherin gamma-A1
Mouse
Protein G purified
Primary
Detects approx 100kDa. Cross-reacts with all Gamma-protocadherin-A, -B and -C proteins.
Store at 4C in the dark.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.