Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCDHGA1 Antibody (S159-5), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242207R
Description
PCDHGA1 Monoclonal specifically detects PCDHGA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
PCDHGA1 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PCDHGA1 | |
Fusion protein corresponding to amino acids 808-931 (C-terminal cytoplasmic constant domain) of mouse Protocadherin-gamma-A1 that is shared by all 22 Gamma-protocadherins (A subfamily amino acids ∼807-930, B subfamily amino acids ∼789-912 and C subfamily amino acids ∼818-941). 99% identity with human (123/124 amino acids). | |
0.1 mL | |
56114 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S159-5 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
MGC138287, MGC138289, PCDH-GAMMA-A1, protocadherin gamma subfamily A, 1, protocadherin gamma-A1 | |
Mouse | |
Protein G purified | |
Primary | |
Detects approx 100kDa. Cross-reacts with all Gamma-protocadherin-A, -B and -C proteins. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction