Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDHGA1 Monoclonal Antibody (4C9)
GREENER_CHOICE

Catalog No. PIMA524552
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524552 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524552 Supplier Invitrogen™ Supplier No. MA524552
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: AFTQAQYHIN VPENVPLGTQ LLMVNATDPD EGANGEVTYS FHNVDHRVAQ IFRLDSYTGE ISNKEPLDFE EYKMYSMEVQ AQDGAGLMAK VKVLIKVLDV.

This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDHGA1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4C9
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PCDHGA1
Gene Accession No. Q9Y5H4
Gene Alias PCDHGA1; PCDH-GAMMA-A1; protocadherin gamma a1; protocadherin gamma subfamily A, 1; protocadherin gamma-A1
Gene Symbols PCDHGA1
Host Species Mouse
Immunogen PCDHGA1 (NP_061735, 241 a.a. ~ 340 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56114
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.