Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pcdhga1, Mouse anti-Mouse, R-PE, Clone: S159-5, Abnova™

Catalog No. 89338512 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-338-512 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-338-512 Supplier Abnova Corporation Supplier No. MAB18587
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen Pcdhga1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone S159-5
Conjugate R-PE
Dilution Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
Gene protocadherin gamma subfamily A, 1
Gene Alias -
Gene Symbols Pcdhga1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 808-931 at C-terminus of mouse Pcdhga1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93709
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.