Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDHGA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552603
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552603 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552603 Supplier Invitrogen™ Supplier No. PA552603
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PNTDWRFSQA QRPGTSGSQN GDDTGTWPNN QFDTEMLQAM ILASASEAAD GSSTLGGGAG TMGLSARYGP QFTLQHVPDY RQNVYIPGSN ATLTNAAGKR DGKAPAGGNG NKKKSGKKEK Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 89%.

This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDHGA2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCDHGA2
Gene Accession No. Q9Y5H1
Gene Alias mKIAA0588; Pcdhga2; PCDH-GAMMA-A2; protocadherin gamma subfamily A, 2; protocadherin gamma-A2
Gene Symbols PCDHGA2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PCDHGA1. Recombinant protein control fragment (Product #RP-104307).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 498846, 56113, 93710
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.