Learn More
Invitrogen™ PCDHGA10 Monoclonal Antibody (3A7)

Description
Peptide Sequence: SDGGDPLRSG TVLVSVTVFD ANDNAPVFTL PEYRVSVPEN LPVGTQLLTV TATDRDEGAN GEVTYSFRKL PDTQLLKFQL NKYTGEIKIS ENLDYEET.
Specifications
Specifications
| Antigen | PCDHGA10 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3A7 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | PCDHGA10 |
| Gene Accession No. | Q9Y5H3 |
| Gene Alias | PCDHGA10; PCDH-gamma-A10; protocadherin gamma subfamily A, 10; protocadherin gamma-A10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.