Learn More
Invitrogen™ PCDHGA2 Monoclonal Antibody (2A7)

Description
Peptide Sequence: SGTSRICVKV LDANDNAPVF TQPEYRISIP ENTLVGTRIL TVTATDADEG YYAQVVYFLE KSPGETSEVF ELKSTSGELT IIKDLDYEDA TFHEIDIEAQ DGPGLLTRA.
Specifications
Specifications
| Antigen | PCDHGA2 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2A7 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | PCDHGA2 |
| Gene Accession No. | Q9Y5H1 |
| Gene Alias | mKIAA0588; Pcdhga2; PCDH-GAMMA-A2; protocadherin gamma subfamily A, 2; protocadherin gamma-A2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.