Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDHGA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571834
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571834 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571834 Supplier Invitrogen™ Supplier No. PA571834
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PCDHGA3 Polyclonal Antibody for Flow, IHC (P), Western Blot

Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintenance of specific neuronal connections in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDHGA3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PCDHGA3
Gene Accession No. Q9Y5H0
Gene Alias Gamma-Protocadherin-A3; PCDG3; PCDH gamma A3; Pcdhga3; PCDH-gamma-A3; protocadherin gamma subfamily A, 3; protocadherin gamma-A3
Gene Symbols PCDHGA3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 284-311 amino acids from the Central region of human PCDHGA3.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56112
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.