Learn More
Invitrogen™ PCDHGA5 Monoclonal Antibody (5H5)

Description
Peptide Sequence: ETVHDLLLTA LDGGDPVLSG TTHIRVTVLD ANDNAPLFTP SEYSVSVPEN IPVGTRLLML TATDPDEGIN GKLTYSFRNE EEKISETFQL DSNLGEISTL.
Specifications
Specifications
| Antigen | PCDHGA5 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 5H5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | PCDHGA5 |
| Gene Accession No. | Q9Y5G8 |
| Gene Alias | cadherin ME3; CDH-GAMMA-A5; ME3; PCDHGA5; PCDH-GAMMA-A5; protocadherin gamma subfamily A, 5; protocadherin gamma-A5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.