Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCDHGB1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | PCDHGB1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15924020
![]() |
Novus Biologicals
NBP15924020UL |
20 μL |
Each for $206.00
|
|
|||||
NBP159240
![]() |
Novus Biologicals
NBP159240 |
100 μL |
Each for $487.50
|
|
|||||
Description
PCDHGB1 Polyclonal specifically detects PCDHGB1 in Human samples. It is validated for Western Blot.Specifications
PCDHGB1 | |
Polyclonal | |
Rabbit | |
Q3SY75 | |
56104 | |
Synthetic peptides corresponding to PCDHGB1(protocadherin gamma subfamily B, 1) The peptide sequence was selected from the N terminal of PCDHGB1. Peptide sequence SPDGSKYPVLLLEKPLDREHQSSHRLILTAMDGGDPPLSGTTHIWIRVTD. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
MGC119466, MGC119467, MGC119469, PCDH-GAMMA-B1, protocadherin gamma subfamily B, 1, protocadherin gamma-B1 | |
PCDHGB1 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title