Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCDHGB7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144374
Description
PCDHGB7 Polyclonal Antibody for ICC/IF, ELISA
Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintenance of specific neuronal connections in the brain.
Specifications
PCDHGB7 | |
Polyclonal | |
Unconjugated | |
PCDHGB7 | |
PCDH-gamma-B7; PCDHGB7; Protocadherin gamma-B7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56099 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunocytochemistry | |
2.695 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q9Y5F8 | |
PCDHGB7 | |
Recombinant Human Protocadherin gamma-B7 protein (719-929AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction