Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCDHGC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567450
Description
Immunogen sequence: YKWKQSRDLYR APVSSLYRTP GPSLHADAVR GGLMSPHLYH QVYLTTDSRR SDPLLKKPGA ASPLASRQNT LRSCDPVFYR QVLGAES Highest antigen sequence identity to the following orthologs - mouse 98%, rat 95%.
This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
Specifications
PCDHGC3 | |
Polyclonal | |
Unconjugated | |
PCDHGC3 | |
Cadherin like 2; cadherin-like 2; PC 43; PC43; PC-43; PCDH 2; PCDH gamma C3; PCDH2; PCDH-GAMMA-C3; Pcdhgb6; PCDHGC 3; PCDHGC3; protocadherin 2; protocadherin 43; protocadherin gamma subfamily C, 3; protocadherin gamma-C3; Protocadherin2; Protocadherin-2; Protocadherin43; protocadherin-43 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5098 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UN70 | |
PCDHGC3 | |
Recombinant Human PCDHGC3. Recombinant protein control fragment (Product #RP-105604). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction