Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCDHGC5 Antibody (OTI2F3), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273270AF594
Description
PCDHGC5 Monoclonal specifically detects PCDHGC5 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PCDHGC5 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
PCDHGC5 | |
Recombinant protein fragment corresponding to amino acids 35-258 of human PCDHGC5 (NP_115783) produced in E.coli. | |
0.1 mL | |
56097.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI2F3 | |
Western Blot | |
MGC138286, MGC138288, PCDH-GAMMA-C5, protocadherin gamma subfamily C, 5, protocadherin gamma-C5 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction