Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDHGC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565666
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565666 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565666 Supplier Invitrogen™ Supplier No. PA565666
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KCLQGNADGDG GGGQCCRRQD SPSPDFYKQS SPNLQVSSDG TLKYMEVTLR PTDSQSHCYR TCFSPASDGS DFTFLRPLSV QQPTALALEP DAIRSRSNTL RERSQQ Highest antigen sequence identity to the following orthologs - mouse 88%, rat 26%.

This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDHGC5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCDHGC5
Gene Accession No. Q9Y5F6
Gene Alias PCDH-gamma-C5; PCDHGC5; protocadherin gamma c5; protocadherin gamma subfamily C, 5; protocadherin gamma-C5
Gene Symbols PCDHGC5
Host Species Rabbit
Immunogen Recombinant Human PCDHGC5. Recombinant protein control fragment (Product #RP-106341).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56097
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.