Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCED1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562248
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562248 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562248 Supplier Invitrogen™ Supplier No. PA562248
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPPPIPGPNP HGQHWGPVVH RGMPRYVPNS PYHVRRMGGP CRQRLRHSER LIHTYKLDRR PPAHS Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

The protein encoded by this gene is a member of the GDSL/SGNH superfamily. Members of this family are hydrolytic enzymes with esterase and lipase activity and broad substrate specificity. This protein belongs to the Pmr5-Cas1p-esterase subfamily in that it contains the catalytic triad comprised of serine, aspartate and histidine and lacks two conserved regions (glycine after strand S2 and GxND motif). A pseudogene of this gene has been identified on the long arm of chromosome 2. Alternative splicing results in multiple transcript variants that encode different protein isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCED1A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCED1A
Gene Accession No. Q9H1Q7
Gene Alias A930025D01Rik; AI480484; bA12M19.1; C20orf81; f113a; FAM113A; family with sequence similarity 113, member A; Pced1a; PC-esterase domain containing 1A; PC-esterase domain-containing protein 1A; Protein FAM113A; RGD1308836; Sarcoma antigen NY-SAR-23
Gene Symbols PCED1A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PCED1A. Recombinant protein control fragment (Product #RP-99429).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64773
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.