Learn More
Invitrogen™ PCF11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110080
Description
Immunogen sequence: PKAYPDNHLS QVDVNELFSK LLKTGILKLS QTDSATTQVS EVTAQPPPEE EEDQNEDQDV PDLTNFTVEE LKQRYDSVIN RLYTGIQCYS CGMRFTTSQT DVYADHLDWH YRQNRTEKDV SRKVTHRRWY YSLTDWIEFE EIADLEERAK SQFFEKVHEE VVLKTQEAAK EKEFQSVPAG PAGAVESCEI CQEQFEQYWD EEEEEWHLKN AIRVDGKIYH PSCYEDYQNT SSFDCTPSPS KTPVENPLNI MLNIVKNELQ EPCDSPKVKE ERIDTPPACT EESIATPSEI KTENDTVESV.
Component of pre-mRNA cleavage complex II.
Specifications
| PCF11 | |
| Polyclonal | |
| Unconjugated | |
| PCF11 | |
| 2500001H09Rik; 5730417B17Rik; C77803; cleavage and polyadenylation factor subunit homolog (S. cerevisiae); KIAA0824; PCF11; PCF11 cleavage and polyadenylation factor subunit; PCF11, cleavage and polyadenylation factor II subunit, homolog; PCF11, cleavage and polyadenylation factor subunit, homolog; PCF11p homolog; pre-mRNA cleavage complex 2 protein Pcf11; Pre-mRNA cleavage complex II protein Pcf11 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51585, 74737 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.59 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O94913 | |
| PCF11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1256-1555 of human PCF11 (NP_056969.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.