Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCF11 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA565277

Catalog No. PIPA565277


Only null left
Add to cart

Description

Description

Immunogen sequence: GIQCYSCGMRF TTSQTDVYAD HLDWHYRQNR TEKDVSRKVT HRRWYYSLTD WIEFEEIADL EERAKSQFFE KVHEEVVLKT QEAAKEKEFQ SVPAG Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

Component of pre-mRNA cleavage complex II.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PCF11
Polyclonal
Unconjugated
PCF11
2500001H09Rik; 5730417B17Rik; C77803; cleavage and polyadenylation factor subunit homolog (S. cerevisiae); KIAA0824; PCF11; PCF11 cleavage and polyadenylation factor subunit; PCF11, cleavage and polyadenylation factor II subunit, homolog; PCF11, cleavage and polyadenylation factor subunit, homolog; PCF11p homolog; pre-mRNA cleavage complex 2 protein Pcf11; Pre-mRNA cleavage complex II protein Pcf11
Rabbit
Antigen affinity chromatography
RUO
51585
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O94913
PCF11
Recombinant Human PCF11
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only