Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCF11 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734090 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4090 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4090 Supplier Proteintech Group Inc Supplier No. 235401AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is specific to the 173 kDa human PCF11 protein.

Component of pre-mRNA cleavage complex II.

Specifications

Antigen PCF11
Applications Western Blot
Classification Polyclonal
Concentration 0.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PCF11
Gene Accession No. O94913
Gene Alias KIAA0824, PCF11
Gene Symbols PCF11
Host Species Rabbit
Immunogen PCF11 Fusion Protein Ag20263
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51585
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.