Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCGF5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP155283
Description
PCGF5 Polyclonal specifically detects PCGF5 in Human samples. It is validated for Western Blot.Specifications
PCGF5 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
MGC16202, polycomb group ring finger 5, polycomb group RING finger protein 5, ring finger protein (C3HC4 type) 159, RING finger protein 159, RNF159 | |
Rabbit | |
30 kDa | |
100 μL | |
Zinc Finger | |
84333 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q86SE9 | |
PCGF5 | |
Synthetic peptides corresponding to PCGF5(polycomb group ring finger 5) The peptide sequence was selected from the N terminal of PCGF5. Peptide sequence ATQRKHLVKDFNPYITCYICKGYLIKPTTVTECLHTFCKTCIVQHFEDSN. | |
Affinity purified | |
RUO | |
Primary | |
Zebrafish: 79%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction