Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCGF5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140096
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DVLCNGEIMG KDHTMEFIYM TRWRLRGENF RCLNCSASQV CSQDGPLYQS YPMVLQYRPR ID.
Component of a Polycomb group (PcG) multiprotein PRC1-like complex, a complex class required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility (PubMed:26151332). Within the PRC1-like complex, regulates RNF2 ubiquitin ligase activity (PubMed:26151332). Plays a redundant role with PCGF3 as part of a PRC1-like complex that mediates monoubiquitination of histone H2A 'Lys-119' on the X chromosome and is required for normal silencing of one copy of the X chromosome in XX females. [UniProt].
Specifications
PCGF5 | |
Polyclonal | |
Unconjugated | |
PCGF5 | |
0610009F02Rik; 1110054A01Rik; 5830406C17Rik; 5830443C21Rik; 9530023M17Rik; AI324127; Pcgf5; polycomb group ring finger 5; polycomb group RING finger protein 5; polycomb group RING finger protein 5-like protein; ring finger protein (C3HC4 type) 159; RING finger protein 159; Rnf159 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84333 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86SE9 | |
PCGF5 | |
Recombinant protein corresponding to Human PCGF5. Recombinant protein control fragment (Product #RP-108965). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction