Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PCI 29732
GREENER_CHOICE

Catalog No. 501250R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-125-0R 50 mg
50-121-0R 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-125-0R Supplier Tocris Bioscience™ Supplier No. 5012/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent BTK inhibitor

PCI 29732 is a potent BTK inhibitor (IC50 = 0.3 nM). Blocks B cell antigen receptor (BCR)-mediated gene expression in CD20+ B cells.

Specifications

Quantity 50 mg
Product Type PCI 29732
Purity >99%
Molecular Weight (g/mol) 371.44
Inhibitors PCI 29732

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.