Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCID1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556626
Description
Immunogen sequence: GGLHVDLAQI IEACDVCLKE DDKDVESVMN SVVSLLLILE PDKQEALIES LCEKLVKFRE GERPSLRLQL LSNLFHGMDK NTPVRYTVYC SLIKVAASCG AIQYIPTELD QVRKWISDWN LTTEKKHTLL RLLYEALVDC KKS Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
This gene encodes a protein that is part of the eurkaryotic translation initiation factor 3 complete (eIF-3) required for protein synthesis. Elevated levels of the encoded protein are present in cancer cell lines. Inactivation of the encoded protein has been shown to interfere with translation of herpes virus mRNAs by preventing the association of mRNAs with the ribosomes. A pseudogene of this gene is located on the X chromosome.
Specifications
PCID1 | |
Polyclonal | |
Unconjugated | |
EIF3M | |
B5; B5 receptor; dendritic cell protein; dendritic cell protein GA17; eIF3m; Eukaryotic translation initiation factor 3 subunit M; eukaryotic translation initiation factor 3, subunit M; fetal lung protein B5; FLJ29030; GA17; HFLB5; hFL-B5; PCI domain containing 1 (herpesvirus entry mediator); PCI domain-containing protein 1; PCID1; PNAS-125; RGD1565840; Tango7; transport and golgi organization 7 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10480 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7L2H7 | |
EIF3M | |
Recombinant protein corresponding to Human PCID1. Recombinant protein control fragment (Product #RP-101061). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction