Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCID2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567269
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567269 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567269 Supplier Invitrogen™ Supplier No. PA567269
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VCASDTRAGIE DSKKWGMLFL VNQLFKIYFK INKLHLCKPL IRAIDSSNLK DDYSTAQRVT YKYYVGRKAM FDS Highest antigen sequence identity to the following orthologs - mouse 99%, rat 100%.

Required for B-cell survival through the regulation of the expression of cell-cycle checkpoint MAD2L1 protein during B cell differentiation. Component of the TREX-2 complex (transcription and export complex 2), composed of at least ENY2, GANP, PCID2, DSS1, and either centrin CETN2 or CETN3 (PubMed:22307388). The TREX-2 complex functions in docking export-competent ribonucleoprotein particles (mRNPs) to the nuclear entrance of the nuclear pore complex (nuclear basket). TREX-2 participates in mRNA export and accurate chromatin positioning in the nucleus by tethering genes to the nuclear periphery. Binds and stabilizes BRCA2 and is thus involved in the control of R-loop-associated DNA damage and transcription-associated genomic instability. R-loop accumulation does not increase in PCID2-depleted cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCID2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCID2
Gene Accession No. Q5JVF3
Gene Alias A730042J05Rik; CSN12-like protein; F10; fa11f03; HT004; hypothetical protein LOC617943; ik:tdsubc_1e10; im:6909928; PCI domain containing 2; PCI domain-containing protein 2; Pcid2; RGD1307041; RP11-98F14.6; tdsubc_1e10; wu:fa11f03; xx:tdsubc_1e10; zgc:101845
Gene Symbols PCID2
Host Species Rabbit
Immunogen Recombinant Human PCID2. Recombinant protein control fragment (Product #RP-107905).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55795
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.