Learn More
PCK1 Mouse anti-Mouse, Unconjugated, Clone: 2F11B5, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | PCK1 |
---|---|
Clone | 2F11B5 |
Concentration | 760 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-9059
![]() ![]() |
Proteintech Group Inc
668621IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The PCK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PCK1. This antibody recognizes mouse, rat, pig, human antigen. The PCK1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
PCK1 | |
760 μg/mL | |
Monoclonal | |
Mouse | |
5105 | |
Primary |
2F11B5 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Mouse, Rat, Pig, Human | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.