Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCK2 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1002422
Description
PCK2 Polyclonal antibody specifically detects PCK2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA.Specifications
PCK2 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
EC 4.1.1.32, PE, PEPCK, PEPCK2PEP carboxykinase, PEPCK-M, phosphoenolpyruvate carboxykinase [GTP], mitochondrial, phosphoenolpyruvate carboxykinase 2 (mitochondrial), Phosphoenolpyruvate carboxylase, phosphopyruvate carboxylase | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize isoform 1 (NP_004554) only, no cross-reactivity with PCK1. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA | |
0.5 mg/mL | |
Western Blot 1 - 3 ug/mL, Flow Cytometry 10 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA Detection limit 1:4000 | |
NP_004554.2 | |
PCK2 | |
Peptide with sequence KPWKPGDKEPCAH corresponding to internal region according to NP_004554.2. | |
0.1 mg | |
Lipid and Metabolism | |
5106 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction