Learn More
Invitrogen™ PCK2 Monoclonal Antibody (3D3D9)

Description
This antibody has been tested in indirect ELISA.
Specifications
Specifications
| Antigen | PCK2 |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3D3D9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | PCK2 |
| Gene Accession No. | Q16822 |
| Gene Alias | 1810010O14Rik; 9130022B02Rik; HGNC:8725; PCK2; PEP carboxykinase; PEPCK; PEPCK2; PEPCK-M; Phosphoenolpyruvate carboxykinase [GTP], mitochondrial; phosphoenolpyruvate carboxykinase 2 (mitochondrial); phosphoenolpyruvate carboxykinase 2, mitochondrial; phosphoenolpyruvate carboxylase; phosphopyruvate carboxylase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.