Learn More
Invitrogen™ PCLO Monoclonal Antibody (6H9-B6)

Description
1 μg/mL of MA5-27629 was sufficient for detection of Piccolo in 20 μg of rat brain tissue lysate by colorimetric immunoblot analysis using goat IgG:HRP as the secondary antibody. Detects approximately 550kDa. No cross-reactivity to Bassoon.
Specifications
Specifications
| Antigen | PCLO |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 6H9-B6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 |
| Gene | PCLO |
| Gene Accession No. | Q9JKS6, Q9QYX7, Q9Y6V0 |
| Gene Alias | Acz; Aczonin; brain-derived HLMN protein; KIAA0559; LOW QUALITY PROTEIN: protein piccolo; multidomain presynaptic cytomatrix protein; multidomain presynaptic cytomatrix protein Piccolo; PCH3; PCLO; piccolo (presynaptic cytomatrix protein); piccolo presynaptic cytomatrix protein; Pico; presynaptic cytomatrix protein; protein piccolo |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.