Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCLO Monoclonal Antibody (PCLO-01)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA528620
Description
This antibody recognizes PCLO (Piccolo), a more than 400 kDa multidomain protein expressed mainly in the presynaptic cytoplasmatic matrix of the neurons.
The protein encoded by the PCLO gene is part of the presynaptic cytoskeletal matrix, which is involved in establishing active synaptic zones and in synaptic vesicle trafficking. Variations in this gene have been associated with bipolar disorder and major depressive disorder. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
PCLO | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Q9Y6V0 | |
PCLO | |
Human recombinant PCLO protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
PCLO-01 | |
Unconjugated | |
PCLO | |
Acz; Aczonin; brain-derived HLMN protein; KIAA0559; LOW QUALITY PROTEIN: protein piccolo; multidomain presynaptic cytomatrix protein; multidomain presynaptic cytomatrix protein Piccolo; PCH3; PCLO; piccolo (presynaptic cytomatrix protein); piccolo presynaptic cytomatrix protein; Pico; presynaptic cytomatrix protein; protein piccolo | |
Mouse | |
Protein A | |
RUO | |
27445 | |
4°C, do not freeze | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction