Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pclo Mouse anti-Human, Mouse, Rat, APC, Clone:6H9-B6, Abnova™

Catalog No. 89333703 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-703 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-703 Supplier Abnova Corporation Supplier No. MAB16973
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full length recombinant mouse Pclo.

Specifications

Antigen Pclo
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 6H9-B6
Conjugate APC
Description piccolo (presynaptic cytomatrix protein)
Dilution ELISA
Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Pclo
Gene Alias Acz, Pico, mKIAA0559
Host Species Mouse
Immunogen Recombinant protein corresponding to full length mouse Pclo.
Quantity 100 μg
Gene ID (Entrez) 26875
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.