Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCM1 Antibody (CL0206), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00
Specifications
Antigen | PCM1 |
---|---|
Clone | CL0206 |
Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
PCM1 Monoclonal antibody specifically detects PCM1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
PCM1 | |
Western Blot 1 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
hPCM-1, pericentriolar material 1, pericentriolar material 1 protein, PTC4PCM-1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: TIYSEVATLISQNESRPHFLIELFHELQLLNTDYLRQRALYALQDIVSRHISESHEKGENVKSVNSGTWIASNSELTPSESLATTDDETFEKNFE | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
CL0206 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Mitotic Regulators | |
PBS (pH 7.2), 40% Glycerol | |
5108 | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title