Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCM1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB10058828
Description
PCM1 Polyclonal specifically detects PCM1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| PCM1 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| hPCM-1, pericentriolar material 1, pericentriolar material 1 protein, PTC4PCM-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot, Immunoprecipitation 2-5 mcg/mg lysate | |
| Q15154 | |
| PCM1 | |
| The immunogen recognized by this antibody maps to a region between residue 1925 and 1975 of human pericentriolar material 1 using the numbering given in entry NP_006188.2 (GeneID 5108). | |
| 0.1 mL | |
| Mitotic Regulators | |
| 5108 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction