Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCM1 Monoclonal Antibody (CL0206)
GREENER_CHOICE

Catalog No. PIMA524589
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524589 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524589 Supplier Invitrogen™ Supplier No. MA524589
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: TIYSEVATLI SQNESRPHFL IELFHELQLL NTDYLRQRAL YALQDIVSRH ISESHEKGEN VKSVNSGTWI ASNSELTPSE SLATTDDETF EKNFE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%. Binds to an epitope located within the peptide sequence RQRALYALQD as determined by overlapping synthetic peptides.

PCM-1 is required for centrosome assembly and function. This protein is essential for the correct localization of several centrosomal proteins including CEP250, CETN3, PCNT and NEK2. The protein is required to anchor microtubules to the centrosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCM1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL0206
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCM1
Gene Accession No. Q15154
Gene Alias 2600002H09Rik; 9430077F19Rik; C030044G17Rik; hPCM-1; mPCM-1; Pcm1; PCM-1; pericentriolar material 1; pericentriolar material 1 protein; pericentriolar material 1, PCM1; pericentriolar material gene 1; PTC4; RET/PCM-1
Gene Symbols PCM1
Host Species Mouse
Immunogen Recombinant protein fragment of Human PCM1 (Product #RP-93739)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5108
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.