Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCM1 Polyclonal Antibody

PCM1 Polyclonal Antibody
Supplier: Thermo Scientific PA576769
Description
PCM1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Specifications
PCM1 | |
Polyclonal | |
Unconjugated | |
PCM1 | |
2600002H09Rik; 9430077F19Rik; C030044G17Rik; hPCM-1; mPCM-1; Pcm1; PCM-1; pericentriolar material 1; pericentriolar material 1 protein; pericentriolar material 1, PCM1; pericentriolar material gene 1; PTC4; RET/PCM-1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18536, 5108, 81740 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15154, Q9R0L6 | |
PCM1 | |
Recombinant full length Human PCM1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction