Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCM1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA595822

Catalog No. PIPA595822


Only null left
Add to Cart

Description

Description

Immunogen sequence: MATGGGPFED GMNDQDLPNW SNENVDDRLN NMDWGAQQKK ANRSSEKNKK KFGVESDKRV TNDISPESSP GVGRRRTKTP HTFPHSRYMS QMSVPEQAEL EKLKQRINFS DLDQRSIGSD SQGRATAANN KRQLSENRKP FNFLPMQINT NKSKDASTNP PNRETIGSAQ CKELFASALS NDLLQNCQVS EEDGRGEPAM ESSQIVSRLV QIRDYITKAS SMREDLVEKN ERSANVERLT HLIDHLKEQE KSYMKFLKKI Positive Samples: Raji, U-87MG; Cellular Location: Cytoplasm, Cytoplasmic granule, centriolar satellite, centrosome, cilium basal body, cytoskeleton, microtubule organizing center.

PCM-1 is required for centrosome assembly and function. This protein is essential for the correct localization of several centrosomal proteins including CEP250, CETN3, PCNT and NEK2. The protein is required to anchor microtubules to the centrosome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PCM1
Polyclonal
Unconjugated
PCM1
2600002H09Rik; 9430077F19Rik; C030044G17Rik; hPCM-1; mPCM-1; Pcm1; PCM-1; pericentriolar material 1; pericentriolar material 1 protein; pericentriolar material 1, PCM1; pericentriolar material gene 1; PTC4; RET/PCM-1
Rabbit
Affinity Chromatography
RUO
5108
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot, Immunocytochemistry
2.17 mg/mL
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Q15154
PCM1
Recombinant protein (or fragment).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.