Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554779
Description
Immunogen sequence: TIYSEVATLI SQNESRPHFL IELFHELQLL NTDYLRQRAL YALQDIVSRH ISESHEKGEN VKSVNSGTWI ASNSELTPSE SLATTDDETF EKNFE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.
PCM-1 is required for centrosome assembly and function. This protein is essential for the correct localization of several centrosomal proteins including CEP250, CETN3, PCNT and NEK2. The protein is required to anchor microtubules to the centrosome.
Specifications
PCM1 | |
Polyclonal | |
Unconjugated | |
PCM1 | |
2600002H09Rik; 9430077F19Rik; C030044G17Rik; hPCM-1; mPCM-1; Pcm1; PCM-1; pericentriolar material 1; pericentriolar material 1 protein; pericentriolar material 1, PCM1; pericentriolar material gene 1; PTC4; RET/PCM-1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5108 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.30 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15154 | |
PCM1 | |
Recombinant protein corresponding to Human PCM1. Recombinant protein control fragment (Product #RP-93739). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction