Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCMT1 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297711AF700
Description
PCMT1 Polyclonal specifically detects PCMT1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PCMT1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 700 | |
EC 2.1.1, EC 2.1.1.77, L-isoaspartyl protein carboxyl methyltransferase, L-isoaspartyl/D-aspartyl methyltransferase, PIMT, Protein L-isoaspartyl/D-aspartyl methyltransferase, Protein-beta-aspartate methyltransferase, protein-L-isoaspartate (D-aspartate) O-methyltransferase, protein-L-isoaspartate(D-aspartate) O-methyltransferase | |
Produced in rabbits immunized with E. coli-derived Human PCMT1 fragment. | |
0.1 mL | |
Proteases & Other Enzymes | |
5110 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction