Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCMT1 Monoclonal Antibody (OTI2A9)
GREENER_CHOICE

Catalog No. PIMA527236
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527236 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527236 Supplier Invitrogen™ Supplier No. MA527236
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PCMT1 Monoclonal Antibody for Western Blot, IHC (P)

Three classes of protein carboxyl methyltransferases, distinguished by their methyl-acceptor substrate specificity, have been found in prokaryotic and eukaryotic cells. The type II enzyme catalyzes the transfer of a methyl group from S-adenosyl-L-methionine to the free carboxyl groups of D-aspartyl and L-isoaspartyl residues. These methyl-accepting residues result from the spontaneous deamidation, isomerization, and racemization of normal L-aspartyl and L-asparaginyl residues and represent sites of covalent damage to aging proteins PCMT1 (EC 2.1.1.77) is a protein repair enzyme that initiates the conversion of abnormal D-aspartyl and L-isoaspartyl residues to the normal L-aspartyl form.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCMT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2A9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PCMT1
Gene Accession No. P22061
Gene Alias C79501; L-isoaspartyl protein carboxyl methyltransferase; PCM; PCMT1; PIMT; protein carboxyl methyltransferase; protein L-isoaspartyl/D-aspartyl methyltransferase; Protein-beta-aspartate methyltransferase; protein-L-isoaspartate (D-aspartate) O-methyltransferase; protein-L-isoaspartate (D-aspartate) O-methyltransferase 1; protein-L-isoaspartate(D-aspartate) O-methyltransferase
Gene Symbols PCMT1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 60-285 of PCMT1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5110
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.