Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCMT1 Monoclonal Antibody (OTI4A4)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527235

Catalog No. PIMA527235


Only null left
Add to Cart

Description

Description

PCMT1 Monoclonal Antibody for Western Blot, IHC (P)

Three classes of protein carboxyl methyltransferases, distinguished by their methyl-acceptor substrate specificity, have been found in prokaryotic and eukaryotic cells. The type II enzyme catalyzes the transfer of a methyl group from S-adenosyl-L-methionine to the free carboxyl groups of D-aspartyl and L-isoaspartyl residues. These methyl-accepting residues result from the spontaneous deamidation, isomerization, and racemization of normal L-aspartyl and L-asparaginyl residues and represent sites of covalent damage to aging proteins PCMT1 (EC 2.1.1.77) is a protein repair enzyme that initiates the conversion of abnormal D-aspartyl and L-isoaspartyl residues to the normal L-aspartyl form.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PCMT1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P22061
PCMT1
Human recombinant protein fragment corresponding to amino acids 60-285 of PCMT1 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
OTI4A4
Unconjugated
PCMT1
C79501; L-isoaspartyl protein carboxyl methyltransferase; PCM; PCMT1; PIMT; protein carboxyl methyltransferase; protein L-isoaspartyl/D-aspartyl methyltransferase; Protein-beta-aspartate methyltransferase; protein-L-isoaspartate (D-aspartate) O-methyltransferase; protein-L-isoaspartate (D-aspartate) O-methyltransferase 1; protein-L-isoaspartate(D-aspartate) O-methyltransferase
Mouse
Affinity chromatography
RUO
5110
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.