Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCMT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551872
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551872 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551872 Supplier Invitrogen™ Supplier No. PA551872
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AKALDVGSGS GILTACFARM VGCTGKVIGI DHIKELVDDS VNNVRKDDPT LLSSGRVQLV VGDGRMGYAE EAPYDAIHVG AAAPVVPQAL IDQLKPGGRL ILPVGPAGGN QMLEQYDKLQ DGSIKMKPLM GVIYVPLTDK EKQWS Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

Three classes of protein carboxyl methyltransferases, distinguished by their methyl-acceptor substrate specificity, have been found in prokaryotic and eukaryotic cells. The type II enzyme catalyzes the transfer of a methyl group from S-adenosyl-L-methionine to the free carboxyl groups of D-aspartyl and L-isoaspartyl residues. These methyl-accepting residues result from the spontaneous deamidation, isomerization, and racemization of normal L-aspartyl and L-asparaginyl residues and represent sites of covalent damage to aging proteins PCMT1 (EC 2.1.1.77) is a protein repair enzyme that initiates the conversion of abnormal D-aspartyl and L-isoaspartyl residues to the normal L-aspartyl form.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCMT1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCMT1
Gene Accession No. P22061, P22062, P23506
Gene Alias C79501; L-isoaspartyl protein carboxyl methyltransferase; PCM; PCMT1; PIMT; protein carboxyl methyltransferase; protein L-isoaspartyl/D-aspartyl methyltransferase; Protein-beta-aspartate methyltransferase; protein-L-isoaspartate (D-aspartate) O-methyltransferase; protein-L-isoaspartate (D-aspartate) O-methyltransferase 1; protein-L-isoaspartate(D-aspartate) O-methyltransferase
Gene Symbols PCMT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PCMT1. Recombinant protein control fragment (Product #RP-96782).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18537, 25604, 5110
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.