Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCMTD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543266
Description
Peptide sequence: TGQNTWESKN ILAVSFAPLV QPSKNDNGKP DSVGLPPCAV RNLQDLARIY Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The function remains unknown.
Specifications
PCMTD1 | |
Polyclonal | |
Unconjugated | |
PCMTD1 | |
8430411F12Rik; A030012M09Rik; FLJ10883; LOW QUALITY PROTEIN: protein-L-isoaspartate O-methyltransferase domain-containing protein 1; PCMTD1; protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1; protein-L-isoaspartate O-methyltransferase domain-containing protein 1; RGD1307986 | |
Rabbit | |
Protein A | |
RUO | |
115294 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96MG8 | |
PCMTD1 | |
Synthetic peptide directed towards the middle region of human PCMTD1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction