Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCMTD2 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306361B
Description
PCMTD2 Polyclonal specifically detects PCMTD2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PCMTD2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C20orf36, chromosome 20 open reading frame 36, FLJ10883, KIAA0835, KIAA1050, protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 2, protein-L-isoaspartate O-methyltransferase domain-containing protein 2 | |
Produced in rabbits immunized with E. coli-derived Human PCMTD2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Biotin | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
55251 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction